accessibility menu, dialog, popup

UserName

CAS RN 22712-18-1

H3COONNH3C

CAS RN 22712-18-1

IUPAC Name: ethyl 2-(3-methylquinoxalin-2-yl)acetate
Synonyms: ethyl 2-(3-methylquinoxalin-2-yl)acetate
Molecular Weight (g/mol): 230.27
Molecular Formula: C13H14N2O2
InChi Key: ZTJBBLDJQXJLCI-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=C(C)N=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results