missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methoxyphenylacetate, 98%

Product Code. 11497357
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497357 25 g 25g
11487357 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497357 Supplier Thermo Scientific Alfa Aesar Supplier No. A15070.14
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 35553-92-5 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35553-92-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00026897
InChI Key XXVVNHCWPHMLEZ-UHFFFAOYSA-N
Synonym ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester
PubChem CID 2774971
IUPAC Name ethyl 2-(3-methoxyphenyl)acetate
SMILES CCOC(=O)CC1=CC(=CC=C1)OC

Specifications

Boiling Point 109°C to 110°C (1 mmHg)
Quantity 25 g
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material Ethyl 3-methoxyphenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.