missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-mercaptopropionate, 98%

Product Code. 15254634 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15254634 25 g Glass bottle 25g
15244634 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254634 Supplier Thermo Scientific Acros Supplier No. 137410250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5466-06-8 | ClH4NO | 69.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5466-06-8
IUPAC Name ethyl 3-sulfanylpropanoate

Specifications

Color Colorless to Yellow
Density 1.0590g/mL
Boiling Point 75.0°C to 76.0°C (10.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H10O2S
Linear Formula HSCH2CH2CO2C2H5
Refractive Index 1.4560 to 1.4580
Quantity 25 g
Beilstein 03,555
Specific Gravity 1.059
Synonym ethyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, ethyl ester, 3-mercaptopropanoic acid ethyl ester, 3-mercaptopropionic acid ethyl ester, ethyl 3-mercaptopropanoate, ethyl beta-mercaptopropionate, unii-22gqz8p70d, propionic acid, 3-mercapto-, ethyl ester, ethyl 3-mercaptopropanoic acid, ethyl .beta.-mercaptopropionate
InChI Key CJQWLNNCQIHKHP-UHFFFAOYSA-N
SMILES CCOC(=O)CCS
Molecular Weight (g/mol) 134.2
PubChem CID 21625
Formula Weight 134.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3-mercaptopropionate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-mercaptopropionate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.