missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-furoate, 99%, Thermo Scientific Chemicals

Product Code. 10535301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10535301 1 g Glass bottle 1g
10013360 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10535301 Supplier Thermo Scientific Acros Supplier No. 269030010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-98-2
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00005348
InChI Key LOFDXZJSDVCYAS-UHFFFAOYSA-N
Synonym ethyl 3-furoate, ethyl 3-furancarboxylate, unii-i8q0yn919k, 3-furancarboxylic acid, ethyl ester, 3-furancarboxylic acid ethyl ester, ethyl 3furoate, ethyl3-furancarboxylate, ethyl3-furoate, pubchem19999, acmc-1bczk
PubChem CID 69201
IUPAC Name ethyl furan-3-carboxylate
SMILES CCOC(=O)C1=COC=C1

Specifications

Density 1.1100g/mL
Flash Point 59°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.462
Quantity 1 g
Beilstein 18,4052
Specific Gravity 1.11
Formula Weight 140.14
Percent Purity 99%
Chemical Name or Material Ethyl 3-furoate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.