missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-formylbenzoate, 98%

Product Code. 11419690
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419690 250 mg 250mg
11429690 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419690 Supplier Thermo Scientific Alfa Aesar Supplier No. H50945.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 33745-47-0 | C10H10O3 | 178.187 g/mol

Ethyl 3-formylbenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-formylbenzoate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33745-47-0
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD08064275
InChI Key XZYGRRSTJVFZLS-UHFFFAOYSA-N
Synonym 3-formylbenzoic acid ethyl ester, 3-formyl-benzoic acid ethyl ester, 3-carbethoxybenzaldehyde, ethyl 3-methanoylbenzoate, acmc-20an34, 3-ethoxycarbonylbenzaldehyde
PubChem CID 11084437
IUPAC Name ethyl 3-formylbenzoate
SMILES CCOC(=O)C1=CC=CC(=C1)C=O

Specifications

Quantity 250 mg
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material Ethyl 3-formylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.