missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylate hydrochloride, 99%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Specifications
Melting Point | 129°C to 132°C |
Color | White to Beige |
Assay Percent Range | 98.5% min. (Argentometry) |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol |
Formula Weight | 217.69 |
Percent Purity | 99% |
Quantity | 250 mg |
Physical Form | Fine Crystalline Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'