missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Ethyoxypropionate, TRC

Product Code. 30375955
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375955 500 mL
30374517 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30375955 Supplier Toronto Research Chemicals Supplier No. TRCE917835500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763-69-9
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.18
Synonym Ethoxypropionic Acid Ethyl Ester, Ektapro EEP, Ethyl 3-ethoxypropanoate, Ethyl 3-Ethoxypropionate, Ethyl 4-Oxahexanoate, Ethyl Ethoxypropionate, Ethyl β-Ethoxypropionate, NSC 8870, Sankio EEP, Ucar Ester EEP
SMILES CCOCCC(=O)OCC

Specifications

Quantity 500 mL
InChI Formula InChI=1S/C7H14O3/c1-3-9-6-5-7(8)10-4-2/h3-6H2,1-2H3
Formula Weight 146.0943
Chemical Name or Material Ethyl 3-Ethyoxypropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.