missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-ethyl-5-methylisoxazole-4-carboxylate, 98%, Thermo Scientific™

Product Code. 11390084
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390084 5 g 5g
11300094 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11390084 Supplier Thermo Scientific Alfa Aesar Supplier No. L00526.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53064-41-8
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00014094
InChI Key JDWLDZUUTIPZHV-UHFFFAOYSA-N
Synonym ethyl 3-ethyl-5-methylisoxazole-4-carboxylate, ethyl 3-ethyl-5-methyl-4-isoxazolecarboxylate, 3-ethyl-5-methyl-4-isoxazolecarboxylicacid ethylester, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid ethyl ester, acmc-1b03d, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-, ethyl ester
PubChem CID 3745090
IUPAC Name ethyl 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate
SMILES CCC1=NOC(=C1C(=O)OCC)C

Specifications

Density 1.07
Boiling Point 71°C to 72°C (0.5 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.463
Quantity 5 g
Beilstein 1073370
Formula Weight 183.21
Percent Purity 98%
Chemical Name or Material Ethyl 3-ethyl-5-methylisoxazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.