missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-ethoxypropionate, 99+%

Product Code. 11406946
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406946 1000 mL 1000mL
11416946 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406946 Supplier Thermo Scientific Alfa Aesar Supplier No. B24024.AU
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 763-69-9 | C7H14O3 | 146.186 g/mol

Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.

Solubility
Miscible with most organic solvents. Slightly miscible with cold water.

Notes
Incompatible with strong oxidizing agents, metals and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763-69-9
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.186
MDL Number MFCD00051356
InChI Key BHXIWUJLHYHGSJ-UHFFFAOYSA-N
Synonym ethyl 3-ethoxypropionate, 3-ethoxypropionic acid ethyl ester, propanoic acid, 3-ethoxy-, ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl beta-ethoxypropionate, unii-ec38rsj79j, propionic acid, 3-ethoxy-, ethyl ester, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove
PubChem CID 12989
IUPAC Name ethyl 3-ethoxypropanoate
SMILES CCOCCC(=O)OCC

Specifications

Melting Point -100°C
Density 0.949
Boiling Point 169°C to 171°C
Flash Point 59°C (142°F)
Odor Ester-like
Linear Formula CH3CH2OCH2CH2CO2CH2CH3
Refractive Index 1.4065
Quantity 1000 mL
UN Number UN3272
Solubility Information Miscible with most organic solvents. Slightly miscible with cold water.
Formula Weight 146.19
Percent Purity ≥99%
Chemical Name or Material Ethyl 3-ethoxypropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.