missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-ethoxy-3-iminopropionate hydrochloride, 85+%

Product Code. 10041554 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041554 100 g Glass bottle 100g
10123294 5 g Glass bottle 5g
10041074 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10041554 Supplier Thermo Scientific Acros Supplier No. 399501000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2318-25-4 | C7H14ClNO3 | 195.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2318-25-4
Molecular Formula C7H14ClNO3
Molecular Weight (g/mol) 195.64
MDL Number MFCD00051405
InChI Key HYMXUYQKXCHWDC-UHFFFAOYSA-N
PubChem CID 75345
SMILES [H+].[Cl-].CCOC(=N)CC(=O)OCC

Specifications

Melting Point 108°C to 110°C
Assay Percent Range 85% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3CH2OC(=NH)CH2COOCH2CH3·HCL
Quantity 100 g
Synonym ethyl 3-ethoxy-3-iminopropanoate hydrochloride, ethyl 3-ethoxy-3-iminopropionate hydrochloride, ethyl 3-ethoxy-3-iminopropionate hcl, ethyl carbethoxyacetimidate hydrochloride, 3-ethoxy-3-iminopropanoic acid ethyl ester hydrochloride, propanoic acid, 3-ethoxy-3-imino-, ethyl ester, hydrochloride, ethyl beta-ethoxy-beta-iminopropionate hydrochloride, diethyl malonmonoimidate hydrochloride, ethyl ethoxycarbonylacetimidate hydrochloride, ethyl3-ethoxy-3-iminopropionate hydrochloride
Solubility Information Solubility in water: decomposes
IUPAC Name ethyl 3-ethoxy-3-iminopropanoate;hydrochloride
Formula Weight 195.64
Percent Purity 85+%
Chemical Name or Material Ethyl 3-ethoxy-3-iminopropionate hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-ethoxy-3-iminopropionate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.