missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate, 96%, Thermo Scientific Chemicals

Product Code. 15408725
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408725 10 g 10g
15488715 1 g 1g
15498715 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15408725 Supplier Thermo Scientific Alfa Aesar Supplier No. H33967.09
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 128073-16-5 | Molecular Formula: C9H7ClF3NO2 | Molecular Weight: 253.605 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128073-16-5
Molecular Formula C9H7ClF3NO2
Molecular Weight (g/mol) 253.605
MDL Number MFCD06656414
InChI Key COMQYNZHBCNPNW-UHFFFAOYSA-N
Synonym ethyl 3-chloro-5-trifluoromethyl picolinate, ethyl 3-chloro-5-trifluoromethyl pyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl-, ethyl ester, 3-chloro-5-trifluoromethylpyridine-2-carboxylic acid ethyl ester, ethyl 5-trifluoromethyl-3-chloropyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl picolite, ethyl 3-chloro-5trifluoromethyl-2-pyridinecarboxylate
PubChem CID 22013558
IUPAC Name ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
SMILES CCOC(=O)C1=C(C=C(C=N1)C(F)(F)F)Cl

Specifications

Boiling Point 64°C to 66°C (0.008 mmHg)
Quantity 10 g
Formula Weight 253.6
Percent Purity 96%
Chemical Name or Material Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.