missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromopyruvate, tech. 75%

Product Code. 11350124
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11350124 100 g 100g
11330124 5 g 5g
11340124 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11350124 Supplier Thermo Scientific Alfa Aesar Supplier No. L00582.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 70-23-5 | C5H7BrO3 | 195.01 g/mol

Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide.

Solubility
Difficult to mix.

Notes
Moisture Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-23-5
Molecular Formula C5H7BrO3
Molecular Weight (g/mol) 195.01
MDL Number MFCD00000204
InChI Key VICYTAYPKBLQFB-UHFFFAOYSA-N
Synonym ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester
PubChem CID 66144
IUPAC Name ethyl 3-bromo-2-oxopropanoate
SMILES CCOC(=O)C(=O)CBr

Specifications

Density 1.555
Boiling Point 98°C to 100°C (10 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.469
Quantity 100 g
UN Number UN3265
Beilstein 1760158
Sensitivity Moisture sensitive
Solubility Information Difficult to mix.
Formula Weight 195.02
Percent Purity 75%
Grade Technical
Chemical Name or Material Ethyl 3-bromopyruvate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.