missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Ethyl 3-bromopropionate was used in the preparation of (3?,4?-dimethoxyphenyl)-5-oxopentanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 3-bromopropionate was used in the preparation of (3′,4′-dimethoxyphenyl)-5-oxopentanoic acid.
Solubility
Soluble in water, alcohol, chloroform, benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Bases, Reducing agents
Ethyl 3-bromopropionate was used in the preparation of (3′,4′-dimethoxyphenyl)-5-oxopentanoic acid.
Solubility
Soluble in water, alcohol, chloroform, benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Bases, Reducing agents
Spécification
Density | 1.416 |
Boiling Point | 67°C to 71°C (10 mmHg) |
Flash Point | 79°C (174°F) |
Refractive Index | 1.452 |
Quantity | 25 g |
Beilstein | 1700933 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water,alcohol,chloroform,benzene. |
Formula Weight | 181.03 |
Percent Purity | 99% |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'