missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromobenzoate, 98%

Product Code. 10682593 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10682593 5 g Glass bottle 5g
10572471 25 g Glass bottle 25g
10331574 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10682593 Supplier Thermo Scientific Acros Supplier No. 308540050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 24398-88-7 | C9H9BrO2 | 229.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24398-88-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.08
MDL Number MFCD00013529
InChI Key QAUASTLEZAPQTB-UHFFFAOYSA-N
PubChem CID 90488
IUPAC Name ethyl 3-bromobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)Br

Specifications

Color Colorless
Density 1.4300g/mL
Boiling Point 130°C to 131°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CO2C2H5
Refractive Index 1.5405 to 1.5425
Quantity 5 g
Specific Gravity 1.43
Synonym ethyl3-bromobenzoate, 3-bromobenzoic acid ethyl ester, benzoic acid, 3-bromo-, ethyl ester, ethyl-3-bromobenzoate, benzoic acid, m-bromo-, ethyl ester, m-bromo benzoic acid ethyl ester, m-bromobenzoic acid, ethyl ester, pubchem3941, acmc-209gbv, ethyl 3-bromobenzoate
Formula Weight 229.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3-bromobenzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.