missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromobenzoate, 98+%

Product Code. 11413963
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413963 250 g 250g
11493953 10 g 10g
11403963 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413963 Supplier Thermo Scientific Alfa Aesar Supplier No. A10251.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 24398-88-7 | C9H9BrO2 | 229.073 g/mol

Ethyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4'-methyl-biphenyl-3-carboxylic acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4′-methyl-biphenyl-3-carboxylic acid ethyl ester.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24398-88-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00013529
InChI Key QAUASTLEZAPQTB-UHFFFAOYSA-N
PubChem CID 90488
IUPAC Name ethyl 3-bromobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)Br

Specifications

Density 1.43
Boiling Point 261°C
Flash Point >110°C (230°F)
Refractive Index 1.543
Quantity 250 g
Beilstein 1867121
Synonym ethyl3-bromobenzoate, 3-bromobenzoic acid ethyl ester, benzoic acid, 3-bromo-, ethyl ester, ethyl-3-bromobenzoate, benzoic acid, m-bromo-, ethyl ester, m-bromo benzoic acid ethyl ester, m-bromobenzoic acid, ethyl ester, pubchem3941, acmc-209gbv, ethyl 3-bromobenzoate
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-bromobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.