missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Aminobenzoate Methanesulfonate, TRC

Product Code. 30136849
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136849 50 g 50g
17931585 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136849 Supplier Toronto Research Chemicals Supplier No. TRCE67856550G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-86-2
Molecular Formula C9 H11 N O2 . C H4 O3 S
Molecular Weight (g/mol) 261.29
Synonym Benzoic acid, 3-amino-, ethyl ester, methanesulfonate (9CI), Benzoic acid, m-amino-, ethyl ester, methanesulfonate (7CI,8CI), Ethyl 3-aminobenzoate methanesulfonate, Ethyl m-aminobenzoate methanesulfonate, Finquel, MS 222, Metacain, Metacaine, NSC 93790, TS 222, Tricaine, Tricaine methanesulfonate
IUPAC Name ethyl 3-aminobenzoate;methanesulfonic acid
SMILES CCOC(=O)c1cccc(N)c1.CS(=O)(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H11NO2.CH4O3S/c1-2-12-9(11)7-4-3-5-8(10)6-7;1-5(2,3)4/h3-6H,2,10H2,1H3;1H3,(H,2,3,4)
Formula Weight 261.0671
Chemical Name or Material Tricaine Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.