missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminobenzoate, 99+%

Product Code. 10459410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10459410 5 g Glass bottle 5g
10152910 25 g Glass bottle 25g
10561564 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10459410 Supplier Thermo Scientific Acros Supplier No. 117990050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 582-33-2 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 582-33-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007794
InChI Key ZMCBYSBVJIMENC-UHFFFAOYSA-N
Synonym 3-aminobenzoic acid ethyl ester, m-ethoxycarbonylaniline, 3-ethoxycarbonyl aniline, ethyl m-aminobenzoate, benzoic acid, 3-amino-, ethyl ester, m-aminobenzoic acid, ethyl ester, m-aminobenzoate, metacaine, 3-amino-benzoic acid ethyl ester, benzoic acid, m-amino-, ethyl ester
PubChem CID 11400
ChEBI CHEBI:81494
IUPAC Name ethyl 3-aminobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)N

Specifications

Color Brown to Yellow
Density 1.1070g/mL
Boiling Point 172°C to 175°C (13.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CO2C2H5
Refractive Index 1.5595 to 1.5615
Quantity 5 g
Beilstein 14, 389
Specific Gravity 1.107
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,diethyl ether,soluble in chloroform
Formula Weight 165.19
Percent Purity 99+%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Ethyl 3-aminobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-aminobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.