missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-amino-4,4,4-trifluorocrotonate, TRC

Product Code. 30388234
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388234 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388234 Supplier Toronto Research Chemicals Supplier No. TRCE899680500MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3-amino-4,4,4-trifluorocrotonate
CAS 372-29-2
Molecular Formula C6H8F3NO2
Quantity 500 mg
InChI Formula InChI=1S/C6H8F3NO2/c1-2-12-5(11)3-4(10)6(7,8)9/h3H,2,10H2,1H3/b4-3+
SMILES CCOC(=O)\C=C(/N)\C(F)(F)F
Molecular Weight (g/mol) 183.13
Formula Weight 183.0507
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.