missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-amino-4,4,4-trifluorobutyrate, 95%, Thermo Scientific Chemicals

Product Code. 11313926
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313926 1 g 1g
11323926 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313926 Supplier Thermo Scientific Alfa Aesar Supplier No. L17025.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 170804-18-9 | Molecular Formula: C6H10F3NO2 | Molecular Weight: 185.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170804-18-9
Molecular Formula C6H10F3NO2
Molecular Weight (g/mol) 185.146
MDL Number MFCD01075754
InChI Key KGUOZZWGWBBRRF-UHFFFAOYSA-N
Synonym ethyl 3-amino-4,4,4-trifluorobutyrate, ethyl-3-amino-4,4,4-trifluorobutyrate, acmc-20akzx, ethyl3-amino-4,4,4-trifluorobutyrate, 3-amino-4,4,4-trifluorobutyric acid ethyl ester
PubChem CID 2774311
IUPAC Name ethyl 3-amino-4,4,4-trifluorobutanoate
SMILES CCOC(=O)CC(C(F)(F)F)N

Specifications

Boiling Point 70°C to 71°C (4mmHg)
Refractive Index 1.3831
UN Number UN2735
Beilstein 6136102
Quantity 1 g
Formula Weight 185.15
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material Ethyl 3-amino-4,4,4-trifluorobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.