missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-Dimethylbenzoate, TRC

Product Code. 30342669
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30342669 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342669 Supplier Toronto Research Chemicals Supplier No. TRCB124825100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3,5-Dimethylbenzoate
CAS 21239-29-2
Molecular Formula C11H14O2
Synonym 3,5-Dimethylbenzoic Acid Ethyl Ester
SMILES CCOC(=O)C1=CC(=CC(=C1)C)C
Molecular Weight (g/mol) 178.23
Formula Weight 178.0994 g/mol
Percent Purity >95% (HPLC)
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.