missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate, 98%, Thermo Scientific™

Product Code. 10174251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10174251 5 g Glass Bottle 5g
10164251 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174251 Supplier Thermo Scientific Acros Supplier No. 343760050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2199-44-2
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00010645
InChI Key IZSBSZYFPYIJDI-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, ethyl 3,5-dimethylpyrrole-2-carboxylate, ethyl 3,5-dimethyl-2-pyrrolecarboxylate, 1h-pyrrole-2-carboxylic acid, 3,5-dimethyl-, ethyl ester, 3,5-dimethyl-1h-pyrrole-2-carboxylic acidethyl ester, ethyl 3,5-dimethyl-1h-pyrrole-2-carboxylate, pubchem23984, acmc-1cdc7, 3,5-dimethyl-1h-pyrrole-2-carboxylicacidethylester, ksc496q7t
PubChem CID 137477
IUPAC Name ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=C(C=C(N1)C)C

Specifications

Melting Point 122°C to 126°C
Color Yellow
Boiling Point 135°C to 136°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 167.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Ethyl 3, 5-dimethyl-1H-pyrrole-2-carboxylate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.