missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(5-bromo-2-methoxy-3-pyridyl)acrylate, Thermo Scientific™

Product Code. 11454570
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454570 1 g 1g
11464570 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454570 Supplier Thermo Scientific Alfa Aesar Supplier No. H50120.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1197397-26-4
Molecular Formula C11H12BrNO3
Molecular Weight (g/mol) 286.125
MDL Number MFCD07781177
InChI Key YEQTUBXMMANUMB-SNAWJCMRSA-N
Synonym 3-5-bromo-2-methoxy-pyridin-3-yl-acrylic acid ethyl ester, ethyl 3-5-bromo-2-methoxypyridin-3-yl acrylate, ethyl 3-5-bromo-2-methoxy-3-pyridyl acrylate, e-ethyl 3-5-bromo-2-methoxypyridin-3-yl acrylate, 3-5-bromo-2-methoxy-3-pyridyl acrylic acid ethyl ester, ethyl 2e-3-5-bromo-2-methoxy 3-pyridyl prop-2-enoate, ethyl 2e-3-5-bromo-2-methoxypyridin-3-yl prop-2-enoate
PubChem CID 42553148
IUPAC Name ethyl (E)-3-(5-bromo-2-methoxypyridin-3-yl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC(=CN=C1OC)Br

Spécification

Quantity 1 g
Formula Weight 286.13
Chemical Name or Material Ethyl 3-(5-bromo-2-methoxy-3-pyridyl)acrylate
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.