missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(4-methoxybenzoyl)propionate, 98%

Product Code. 11386409
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386409 25 g 25g
11372137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386409 Supplier Thermo Scientific Alfa Aesar Supplier No. L07989.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 15118-67-9 | C13H16O4 | 236.267 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15118-67-9
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.267
MDL Number MFCD00051784
InChI Key FYUAOZFEVHSJTO-UHFFFAOYSA-N
Synonym ethyl 3-4-methoxybenzoyl propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate, ethyl 4-4-methoxyphenyl-4-oxobutyrate, acmc-1ciqg, ethyl 3-4-methoxybenzoylpropionate, ethyl 3-4-methoxybenzoyl propanoate, ethyl 3-4-methoxybenzoyl-propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate #, gamma-oxo-4-methoxybenzenebutyric acid ethyl ester, 4-4-methoxyphenyl-4-oxobutyric acid, ethyl ester
PubChem CID 585132
IUPAC Name ethyl 4-(4-methoxyphenyl)-4-oxobutanoate
SMILES CCOC(=O)CCC(=O)C1=CC=C(C=C1)OC

Specifications

Melting Point 49°C to 53°C
Quantity 25 g
Beilstein 2120509
Formula Weight 236.27
Percent Purity 98%
Chemical Name or Material Ethyl 3-(4-methoxybenzoyl)propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.