missing translation for 'onlineSavingsMsg'
Läs mer

Ethyl 3-(4-methoxybenzoyl)propionate, 98%

Produktkod. 11372137
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Förpackningsstorlek
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11372137 5 g 5g
11386409 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11372137 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' L07989.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 15118-67-9 | C13H16O4 | 236.267 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 15118-67-9
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.267
MDL Number MFCD00051784
InChI Key FYUAOZFEVHSJTO-UHFFFAOYSA-N
Synonym ethyl 3-4-methoxybenzoyl propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate, ethyl 4-4-methoxyphenyl-4-oxobutyrate, acmc-1ciqg, ethyl 3-4-methoxybenzoylpropionate, ethyl 3-4-methoxybenzoyl propanoate, ethyl 3-4-methoxybenzoyl-propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate #, gamma-oxo-4-methoxybenzenebutyric acid ethyl ester, 4-4-methoxyphenyl-4-oxobutyric acid, ethyl ester
PubChem CID 585132
IUPAC Name ethyl 4-(4-methoxyphenyl)-4-oxobutanoate
SMILES CCOC(=O)CCC(=O)C1=CC=C(C=C1)OC

Specifikationer

Melting Point 49°C to 53°C
Quantity 5 g
Beilstein 2120509
Formula Weight 236.27
Percent Purity 98%
Chemical Name or Material Ethyl 3-(4-methoxybenzoyl)propionate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.