missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4-Dihydroxybenzoate, TRC

Product Code. 30384325
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384325 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384325 Supplier Toronto Research Chemicals Supplier No. TRCE67850050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3,4-Dihydroxybenzoate
CAS 3943-89-3
Molecular Formula C9H10O4
Quantity 50 g
Synonym 3,4-Dihydroxybenzoic Acid Ethyl Ester, Protocatechuic Acid Ethyl Ester, Ethyl Protocatechuate, NSC 619681, NSC 86130
InChI Formula InChI=1S/C9H10O4/c1-2-13-9(12)6-3-4-7(10)8(11)5-6/h3-5,10-11H,2H2,1H3
SMILES CCOC(=O)C1=CC(=C(C=C1)O)O
Molecular Weight (g/mol) 182.17
Formula Weight 182.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.