missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4-dihydroxybenzoate, 98%

Product Code. 11464453
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464453 250 g 250g
11444453 10 g 10g
11454453 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464453 Supplier Thermo Scientific Alfa Aesar Supplier No. A11001.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3943-89-3 | C9H10O4 | 182.18 g/mol

The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.

Solubility
Insoluble in water. Soluble in ethanol.

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3943-89-3
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002199
InChI Key KBPUBCVJHFXPOC-UHFFFAOYSA-N
Synonym ethyl protocatechuate, 3,4-dihydroxybenzoic acid ethyl ester, ethyl-3,4-dihydroxybenzoate, unii-4ygj96wtbg, benzoic acid, 3,4-dihydroxy-, ethyl ester, edhb, protocatechuic acid ethyl ester, 4ygj96wtbg, 3,4-dihydroxy-benzoic acid ethyl ester
PubChem CID 77547
IUPAC Name ethyl 3,4-dihydroxybenzoate
SMILES CCOC(=O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 132°C to 135°C
Quantity 250 g
Beilstein 2097435
Solubility Information Insoluble in water. Soluble in ethanol.
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material Ethyl 3,4-dihydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.