missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3,4-dihydroxybenzoate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.
Solubility
Insoluble in water. Soluble in ethanol.
Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, Strong bases.
The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.
Solubility
Insoluble in water. Soluble in ethanol.
Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, Strong bases.
Specifications
Melting Point | 132°C to 135°C |
Quantity | 50 g |
Beilstein | 2097435 |
Solubility Information | Insoluble in water. Soluble in ethanol. |
Formula Weight | 182.18 |
Percent Purity | 98% |
Chemical Name or Material | Ethyl 3,4-dihydroxybenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'