missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(4-Chlorophenyl)-3-oxopropanoate, TRC

Product Code. 30339475
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339475 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339475 Supplier Toronto Research Chemicals Supplier No. TRCB124820100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Ethyl 3-(4-Chlorophenyl)-3-oxopropanoate
CAS 2881-63-2
Molecular Formula C11H11O3Cl
Quantity 100 mg
InChI Formula InChI=1S/C11H11ClO3/c1-2-15-11(14)7-10(13)8-3-5-9(12)6-4-8/h3-6H,2,7H2,1H3
SMILES CCOC(=O)CC(=O)c1ccc(Cl)cc1
IUPAC Name ethyl 3-(4-chlorophenyl)-3-oxopropanoate
Molecular Weight (g/mol) 226.65
Formula Weight 226.0397
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.