missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(4-bromophenyl)propionate, 95%

Product Code. 15448188
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448188 5 g 5g
15458188 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448188 Supplier Thermo Scientific Alfa Aesar Supplier No. H61633.06
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 40640-98-0 | C11H13BrO2 | 257.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40640-98-0
Molecular Formula C11H13BrO2
Molecular Weight (g/mol) 257.13
MDL Number MFCD07772961
InChI Key STMHGPFYLQOGJD-UHFFFAOYSA-N
PubChem CID 12731927
IUPAC Name ethyl 3-(4-bromophenyl)propanoate
SMILES CCOC(=O)CCC1=CC=C(Br)C=C1

Specifications

Quantity 5 g
Synonym ethyl 3-4-bromophenyl propanoate, 3-4-bromophenyl propionic acid ethyl ester, benzenepropanoic acid, 4-bromo-, ethyl ester, ethyl 3-4-bromophenyl-propionate, 3-4-bromo-phenyl-propionic acid ethyl ester, ksc497s3n, ethyl3-4-bromophenyl propanoate, 4-bromobenzenepropanoic acid ethyl ester, 3-4-bromophenyl propanoic acid ethyl ester, 3-4-bromophenyl-propionic acid ethyl ester
Formula Weight 257.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 3-(4-bromophenyl)propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.