missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4,5-trimethoxybenzoate, 98%

Product Code. 11444624
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444624 100 g 100g
11424624 5 g 5g
11434624 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444624 Supplier Thermo Scientific Alfa Aesar Supplier No. A13199.22
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6178-44-5 | C12H16O5 | 240.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6178-44-5
Molecular Formula C12H16O5
Molecular Weight (g/mol) 240.255
MDL Number MFCD00017525
InChI Key UEOFNBCUGJADBM-UHFFFAOYSA-N
PubChem CID 231162
IUPAC Name ethyl 3,4,5-trimethoxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)OC)OC)OC

Specifications

Quantity 100 g
Synonym 3,4,5-trimethoxybenzoic acid ethyl ester, ethyl 3,4,5-trimethoxy benzoate, ethyltrimethoxy benzoate, acmc-1b7mp, ethyl 3,4,5-trimethoxybenzoate;, benzoic acid,3,4,5-trimethoxy-, ethyl ester, benzoic acid, 3,4,5-trimethoxy-, ethyl ester
Formula Weight 240.26
Percent Purity 0.98
Chemical Name or Material Ethyl 3,4,5-trimethoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.