missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4,5-trihydroxybenzoate, 98%

Product Code. 11464754
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464754 50 g 50g
11454754 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464754 Supplier Thermo Scientific Alfa Aesar Supplier No. A13733.18
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 831-61-8 | C9H10O5 | 198.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 831-61-8
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00016430
InChI Key VFPFQHQNJCMNBZ-UHFFFAOYSA-N
Synonym ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
PubChem CID 13250
ChEBI CHEBI:87247
IUPAC Name ethyl 3,4,5-trihydroxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 148°C to 152°C
Quantity 50 g
Formula Weight 198.18
Percent Purity 0.98
Chemical Name or Material Ethyl 3,4,5-trihydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.