missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]propanoate, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Ethyl 3-[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]propanoate
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]propanoate |
| CAS | 868754-42-1 |
| Analyte or Component Names | Ethyl 3-[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]propanoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Pioglitazone Hydrochloride Imp. E (EP), Pioglitazone Imp. E (EP), Ethyl 3-[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]propanoate, Pioglitazone Hydrochloride Imp. E (Pharmeuropa), Pioglitazone Hydrochloride Impurity E, Pioglitazone Impurity E |
| InChI Formula | InChI=1S/C20H25NO3/c1-3-16-5-9-18(21-15-16)13-14-24-19-10-6-17(7-11-19)8-12-20(22)23-4-2/h5-7,9-11,15H,3-4,8,12-14H2,1-2H3 |
| SMILES | CCOC(=O)CCc1ccc(OCCc2ccc(CC)cn2)cc1 |
| IUPAC Name | ethyl 3-[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]propanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction