missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3-diethoxypropionate, 96%

Product Code. 11347496
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347496 10 g 10g
11357496 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347496 Supplier Thermo Scientific Alfa Aesar Supplier No. L09562.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10601-80-6 | C9H18O4 | 190.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10601-80-6
Molecular Formula C9H18O4
Molecular Weight (g/mol) 190.239
MDL Number MFCD00009865
InChI Key SIALOQYKFQEKOG-UHFFFAOYSA-N
PubChem CID 66389
IUPAC Name ethyl 3,3-diethoxypropanoate
SMILES CCOC(CC(=O)OCC)OCC

Specifications

Density 0.978
Boiling Point 92°C to 93°C (13 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.412
Quantity 10 g
Beilstein 1772110
Synonym ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
Formula Weight 190.24
Percent Purity 96%
Chemical Name or Material Ethyl 3,3-diethoxypropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.