missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3,3-trifluoropyruvate, 98+%

Código de producto. 10505334 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Tamaño de la unidad:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging Tamaño de la unidad
10505334 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10505334 Proveedor Thermo Scientific Acros N.º de proveedor 399010050
 Restricted Item
Estimated Shipment: 09-10-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 13081-18-0 | C5H5F3O3 | 170.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

CAS 13081-18-0
Color Colorless to Yellow
Density 1.2830g/mL
Boiling Point 95°C to 98°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C5H5F3O3
Linear Formula CF3C(O)CO2CH2CH3
MDL Number MFCD00114935
Quantity 5 g
Specific Gravity 1.283
Synonym ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
InChI Key KJHQVUNUOIEYSV-UHFFFAOYSA-N
SMILES CCOC(=O)C(=O)C(F)(F)F
IUPAC Name ethyl 3,3,3-trifluoro-2-oxopropanoate
Molecular Weight (g/mol) 170.09
PubChem CID 2737239
Formula Weight 170.09
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Ethyl 3, 3, 3-trifluoropyruvate
Mostrar más Mostrar menos
compliance-icons compliance-icons
Product Identifier
  • Ethyl 3,3,3-trifluoropyruvate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.