missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2-furyl)-1H-pyrazole-5-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 11415500
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415500 5 g 5g
11496430 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415500 Supplier Thermo Scientific Alfa Aesar Supplier No. H25791.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33545-44-7
Molecular Formula C10H10N2O3
Molecular Weight (g/mol) 206.20
MDL Number MFCD09953454,MFCD06797478
InChI Key GSVBXAZEQDJUOY-UHFFFAOYSA-N
Synonym ethyl 5-2-furyl-1h-pyrazole-3-carboxylate, ethyl 5-furan-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-furan-2-yl-1h-pyrazole-5-carboxylate, ethyl 5-fur-2-yl-1h-pyrazole-3-carboxylate, ethyl 5-2-furyl pyrazole-3-carboxylate, ethyl 5-furan-2-yl-2h-pyrazole-3-carboxylate, aronis23991, ethyl 3-2-furyl pyrazole-5-carboxylate, 1h-pyrazole-3-carboxylicacid, 5-2-furanyl-, ethyl ester
PubChem CID 7127811
IUPAC Name ethyl 3-(furan-2-yl)-1H-pyrazole-5-carboxylate
SMILES CCOC(=O)C1=CC(=NN1)C1=CC=CO1

Specifications

Melting Point 135°C to 137°C
Quantity 5 g
Formula Weight 206.2
Percent Purity 98%
Chemical Name or Material Ethyl 3-(2-furyl)-1H-pyrazole-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.