missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2R,4R)-4-Methylpipecolate, TRC

Product Code. 30386277
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386277 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386277 Supplier Toronto Research Chemicals Supplier No. TRCE9246301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl (2R,4R)-4-Methylpipecolate
CAS 74892-82-3
Molecular Formula C9H17NO2
Quantity 1 g
Synonym (2R-trans)-4-Methyl-2-piperidinecarboxylic Acid Ethyl Ester, Ethyl (2R,4R)-4-Methyl-2-piperidinecarboxylate, (2R,4R)-4-Methyl-2-piperidinecarboxylic Acid Ethyl Ester
InChI Formula InChI=1S/C9H17NO2/c1-3-12-9(11)8-6-7(2)4-5-10-8/h7-8,10H,3-6H2,1-2H3/t7-,8-/m1/s1
SMILES CCOC(=O)[C@H]1C[C@H](C)CCN1
Molecular Weight (g/mol) 171.24
Formula Weight 171.1259
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.