Learn More
Ethyl 2-oxopyrrolidine-3-carboxylate, 95%, Thermo Scientific Chemicals
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Recommended storage temperature: 2 - 8°C Handle and store under inert gas.
Specifications
Specifications
| Melting Point | 71°C to 75°C |
| Quantity | 250 mg |
| Sensitivity | Light sensitive |
| Synonym | 2-oxo-pyrrolidine-3-carboxylic acid ethyl ester, ethyl 2-oxo-pyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylic acid, 2-oxo-, ethyl ester, 2-oxopyrrolidine-3-carboxylic acid ethyl ester, 3-carboethoxy-pyrrolidinone, ethyl2-oxo-pyrrolidine-3-carboxylate, ethyl 2-oxopyrrolidine-3-carboxylate, n-boc-2-oxo-d-beta-proline ethyl ester, 3-pyrrolidinecarboxylicacid, 2-oxo-, ethyl ester |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 157.17 |
| Percent Purity | 95% |
| Chemical Name or Material | Ethyl 2-oxopyrrolidine-3-carboxylate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.