missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxopyrrolidine-3-carboxylate, 95%, Thermo Scientific Chemicals

Product Code. 15454298
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454298 1 g 1g
15464298 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454298 Supplier Thermo Scientific Alfa Aesar Supplier No. H57906.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36821-26-8 | Molecular Formula: C7H11NO3 | Molecular Weight: 157.169 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Recommended storage temperature: 2 - 8°C Handle and store under inert gas.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36821-26-8
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.169
MDL Number MFCD06796534
InChI Key FHHMWJMUMOYGQW-UHFFFAOYSA-N
PubChem CID 12699000
IUPAC Name ethyl 2-oxopyrrolidine-3-carboxylate
SMILES CCOC(=O)C1CCNC1=O

Specifications

Melting Point 71°C to 75°C
Quantity 1 g
Sensitivity Light sensitive
Synonym 2-oxo-pyrrolidine-3-carboxylic acid ethyl ester, ethyl 2-oxo-pyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylic acid, 2-oxo-, ethyl ester, 2-oxopyrrolidine-3-carboxylic acid ethyl ester, 3-carboethoxy-pyrrolidinone, ethyl2-oxo-pyrrolidine-3-carboxylate, ethyl 2-oxopyrrolidine-3-carboxylate, n-boc-2-oxo-d-beta-proline ethyl ester, 3-pyrrolidinecarboxylicacid, 2-oxo-, ethyl ester
Solubility Information Slightly soluble in water.
Formula Weight 157.17
Percent Purity 95%
Chemical Name or Material Ethyl 2-oxopyrrolidine-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.