missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxocyclopentylacetate, 95%, Thermo Scientific™

Product Code. 15418148
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418148 25 g 25g
15408148 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418148 Supplier Thermo Scientific Alfa Aesar Supplier No. H61589.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 2-oxocyclopentylacetate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20826-94-2
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00044715
InChI Key PJMKFKUFBDXYEC-SSDOTTSWSA-N
Synonym ethyl 2-oxocyclopentylacetate, ethyl 2-2-oxocyclopentyl acetate, ethyl 2-oxocyclopentaneacetate, ethyl 2-oxocyclopentyl acetate, 2-oxocyclopentaneacetic acid ethyl ester, cyclopentaneacetic acid, 2-oxo-, ethyl ester, pubchem23864, ksc201q9p, ethyl 2-oxo-cyclopentyl acetate, ehtyl 2-2-oxocyclopentyl acetate
PubChem CID 2723663
IUPAC Name ethyl 2-(2-oxocyclopentyl)acetate
SMILES CCOC(=O)C[C@H]1CCCC1=O

Specifications

Refractive Index 1.4517
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 170.21
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 2-oxocyclopentylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.