missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxocyclopentanecarboxylate, 97+%

Product Code. 11351617
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11351617 100 g 100g
11341617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351617 Supplier Thermo Scientific Alfa Aesar Supplier No. L04528.22
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 611-10-9 | C8H12O3 | 156.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-10-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00001412
InChI Key JHZPNBKZPAWCJD-UHFFFAOYSA-N
Synonym ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate
PubChem CID 69136
IUPAC Name ethyl 2-oxocyclopentane-1-carboxylate
SMILES CCOC(=O)C1CCCC1=O

Specifications

Density 1.079
Boiling Point 225°C to 228°C
Flash Point 77°C (170°F)
Refractive Index 1.452
Quantity 100 g
Beilstein 387787
Formula Weight 156.18
Percent Purity ≥97%
Chemical Name or Material Ethyl 2-oxocyclopentanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.