missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Oxo-4-phenylbutyrate-d5, TRC

Product Code. 30380118
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380118 5 mg 5mg
30377518 100 mg -
30384428 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30380118 Supplier Toronto Research Chemicals Supplier No. TRCE9253875MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189911-53-2
Molecular Formula C12 2H5 H9 O3
Molecular Weight (g/mol) 211.27
Synonym Ethyl 2-oxo-4-(2,3,4,5,6-pentadeuteriophenyl)butanoate, Ethyl 2-Oxo-4-phenylbutanoate-D5 (phenyl-D5), Ethyl 2-Keto-4-phenyl-d5-butanoate, Ethyl 2-Oxo-4-phenylbutyrate-d5, Ethyl 2-Oxo-4-(phenyl-d5)butanoate, 2-Oxo-4-(phenyl-d5)butanoic acid ethyl ester
IUPAC Name ethyl 2-oxo-4-(2,3,4,5,6-pentadeuteriophenyl)butanoate
SMILES [2H]c1c([2H])c([2H])c(CCC(=O)C(=O)OCC)c([2H])c1[2H]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H14O3/c1-2-15-12(14)11(13)9-8-10-6-4-3-5-7-10/h3-7H,2,8-9H2,1H3/i3D,4D,5D,6D,7D
Formula Weight 211.1257
Chemical Name or Material Ethyl 2-Oxo-4-phenylbutyrate d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.