missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-octynoate, 98%

Product Code. p-7024680
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498647 5 g 5g
11408657 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498647 Supplier Thermo Scientific Alfa Aesar Supplier No. A19784.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10519-20-7 | C10H16O2 | 168.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10519-20-7
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00015286
InChI Key QPMDWYXUSMRVKT-UHFFFAOYSA-N
Synonym ethyl 2-octynoate, 2-octynoic acid, ethyl ester, 2-octynoic acid ethyl ester, ethyl 2-octynate, ethyl heptynecarbonate, unii-xth780u0n3, ethyl heptine carbonate, ethyl n-oct-2-ynoate, ethyl heptyne carboxylate, 2-octynoic acid, ethylester
PubChem CID 66338
IUPAC Name ethyl oct-2-ynoate
SMILES CCCCCC#CC(=O)OCC

Specifications

Density 0.91
Boiling Point 85°C (6 mmHg)
Flash Point 99°C (210°F)
Linear Formula CH3(CH2)4C≡CCO2CH2CH3
Refractive Index 1.447
Quantity 5 g
Beilstein 1759822
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material Ethyl 2-octynoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.