missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(methylsulfonyl)acetate, 97%, Thermo Scientific™

Product Code. 11975781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11975781 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11975781 Supplier Thermo Scientific Acros Supplier No. 443200050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4455-15-6
Color Colorless to Yellow
Density 1.2590g/mL
Elemental Analysis 34.69 to 37.86% C, 5.82 to 6.31% H
Boiling Point 111°C to 113°C (0.2 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C5H10O4S
Quantity 5 g
Specific Gravity 1.259
Synonym ethyl methanesulfonylacetate, ethyl 2-methylsulfonyl acetate, ethyl methylsulfonylacetate, ethyl methylsulphonyl acetate, ethyl methanesulphonylacetate, ethyl methylsulfonyl acetate, ethyl 2-methanesulfonylacetate, acetic acid, methylsulfonyl-, ethyl ester, ethylmethanesulfonylacetate
InChI Key OCCWQCYBCZADCE-UHFFFAOYSA-N
SMILES CCOC(=O)CS(=O)(=O)C
IUPAC Name ethyl 2-methylsulfonylacetate
Molecular Weight (g/mol) 166.2
PubChem CID 78201
Formula Weight 166.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 2-(methylsulfonyl)acetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-(methylsulfonyl)acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.