missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylnicotinate, 97%

Product Code. 11362076
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362076 25 g 25g
11352076 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362076 Supplier Thermo Scientific Alfa Aesar Supplier No. L03256.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1721-26-2 | C9H11NO2 | 165.19 g/mol

Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1721-26-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00006336
InChI Key TZORNSWZUZDUCU-UHFFFAOYSA-N
Synonym ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727
PubChem CID 74401
IUPAC Name ethyl 2-methylpyridine-3-carboxylate
SMILES CCOC(=O)C1=CC=CN=C1C

Specifications

Density 1.07
Boiling Point 107°C to 108°C (13 mmHg)
Flash Point 102°C (215°F)
Odor Recognizable
Refractive Index 1.505
Quantity 25 g
Beilstein 118200
Solubility Information Slightly soluble in water.
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material Ethyl 2-methylnicotinate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.