missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylacetoacetate, 95%

Product Code. 10286051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10286051 25 g Glass bottle 25g
10337901 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10286051 Supplier Thermo Scientific Acros Supplier No. 326110250
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 609-14-3 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-14-3
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009164
InChI Key FNENWZWNOPCZGK-UHFFFAOYSA-N
Synonym ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
PubChem CID 701
IUPAC Name ethyl 2-methyl-3-oxobutanoate
SMILES CCOC(=O)C(C)C(=O)C

Specifications

Melting Point -45°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 187°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH(CH3)CO2C2H5
Refractive Index 1.4170 to 1.419
Quantity 25 g
Beilstein 03, 679
Specific Gravity 1.01
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents
Formula Weight 144.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 2-methylacetoacetate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.