missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methoxybenzoate, 98+%, Thermo Scientific™

Product Code. 11493654
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493654 50 g 50g
11403664 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493654 Supplier Thermo Scientific Alfa Aesar Supplier No. A11287.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7335-26-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00015148
InChI Key FNODWEPAWIJGPM-UHFFFAOYSA-N
Synonym ethyl o-anisate, benzoic acid, 2-methoxy-, ethyl ester, ethyl ortho-anisate, 2-methoxybenzoic acid ethyl ester, ethyl o-methoxybenzoate, o-anisic acid, ethyl ester, 2-methoxy-benzoic acid ethyl ester, ethyl-2-methoxy benzoate, dsstox_cid_27126, dsstox_rid_82133
PubChem CID 81784
IUPAC Name ethyl 2-methoxybenzoate
SMILES CCOC(=O)C1=CC=CC=C1OC

Specifications

Density 1.112
Boiling Point 127°C to 128°C (11 mmHg)
Refractive Index 1.521
Quantity 50 g
Beilstein 2575515
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.