missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2-Methoxy-2-oxoethyl) Malonate, TRC

Product Code. 30384238
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384238 2.5 g 2.5g
30381505 1 g 1g
30388619 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384238 Supplier Toronto Research Chemicals Supplier No. TRCE5724652.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl (2-Methoxy-2-oxoethyl) Malonate
Analyte or Component Names Ethyl (2-Methoxy-2-oxoethyl) Malonate
Shipping Condition Room Temperature
Molecular Formula C8 H12 O6
InChI Formula InChI=1S/C8H12O6/c1-3-13-6(9)4-7(10)14-5-8(11)12-2/h3-5H2,1-2H3
Solution Type Neat
SMILES CCOC(=O)CC(=O)OCC(=O)OC
IUPAC Name 1-O-ethyl 3-O-(2-methoxy-2-oxoethyl) propanedioate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 204.177
Quantity 2.5 g
Formula Weight 204.063
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.