missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-isothiocyanatopropionate, 97%

Product Code. 11393216
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393216 1 g 1g
11303226 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393216 Supplier Thermo Scientific Alfa Aesar Supplier No. L12532.03
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39574-16-8 | C6H9NO2S | 159.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39574-16-8
Molecular Formula C6H9NO2S
Molecular Weight (g/mol) 159.20
MDL Number MFCD00070636
InChI Key ALJGYASQFZQQJX-UHFFFAOYNA-N
Synonym ethyl 2-isothiocyanatopropionate, ethyl n-thioxomethylene alaninate, propanoic acid, 2-isothiocyanato-, ethyl ester, acmc-20al0z, acmc-20lue7, propanoic acid, 2-isothiocyanato-, ethyl ester, s, ethyl l-2-isothiocyanatopropanoate, alanine, n-carbonothioyl-, ethyl ester
PubChem CID 142361
IUPAC Name ethyl 2-isothiocyanatopropanoate
SMILES CCOC(=O)C(C)N=C=S

Specifications

Boiling Point 63°C to 64°C (1 mmHg)
Refractive Index 1.4925
Quantity 1 g
UN Number UN2922
Beilstein 1723243
Sensitivity Moisture sensitive
Formula Weight 159.2
Percent Purity 97%
Chemical Name or Material Ethyl 2-isothiocyanatopropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.