missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-hydroxyisobutyrate, 98%

Product Code. 11475617
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475617 100 g 100g
11465617 25 g 25g
11485617 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475617 Supplier Thermo Scientific Alfa Aesar Supplier No. B23592.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-55-7 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-55-7
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00004458
InChI Key GFUIDHWFLMPAGY-UHFFFAOYSA-N
Synonym ethyl 2-hydroxyisobutyrate, ethyl 2-hydroxy-2-methylpropionate, ethyl 2-methyllactate, propanoic acid, 2-hydroxy-2-methyl-, ethyl ester, ethyl alpha-hydroxyisobutyrate, 2-methyllactic acid ethyl ester, ethyl, a-hydroxyisobutyrate, lactic acid, 2-methyl-, ethyl ester, unii-s1530mcv3x, ethyl-2-hydroxyisobutyrate
PubChem CID 6653
IUPAC Name ethyl 2-hydroxy-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)O

Specifications

Density 0.98
Boiling Point 149°C to 150°C
Flash Point 44°C (111°F)
Refractive Index 1.408
Quantity 100 g
UN Number UN3272
Beilstein 878308
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material Ethyl 2-hydroxyisobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.