missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-hydroxy-6-methylpyridine-4-carboxylate, 97%

Product Code. 11382949
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382949 1 g 1g
15437725 250 mg 250mg
11392949 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382949 Supplier Thermo Scientific Alfa Aesar Supplier No. H27775.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150190-03-7 | C9H11NO3 | 181.191 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150190-03-7
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.191
MDL Number MFCD09953494
InChI Key WZUAZSMUJJUHMM-UHFFFAOYSA-N
Synonym ethyl 2-hydroxy-6-methylisonicotinate, ethyl 2-hydroxy-6-methylpyridine-4-carboxylate, ethyl 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylate, 4-pyridinecarboxylicacid, 1,2-dihydro-6-methyl-2-oxo-, ethyl ester, acmc-20edky, 2-hydroxy-6-methylpyridine-4-carboxylic acid ethyl ester, 2-methyl-6-oxo-1h-pyridine-4-carboxylic acid ethyl ester, ethyl 2-methyl-6-oxidanylidene-1h-pyridine-4-carboxylate
PubChem CID 12089326
IUPAC Name ethyl 2-methyl-6-oxo-1H-pyridine-4-carboxylate
SMILES CCOC(=O)C1=CC(=O)NC(=C1)C

Specifications

Melting Point 188°C to 190°C
Quantity 1 g
Formula Weight 181.2
Percent Purity 97%
Chemical Name or Material Ethyl 2-hydroxy-6-methylpyridine-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.